siRNA Details
| Gene Symbol: Ndst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031186.3 | NP_112463.2 | 1 | 843 | ||
| GenBank mRNA | AF221095.1 | - | 1 | 842 | ||
| AK141945.1 | - | 2 | 992 | |||
| BC079622.1 | - | 1 | 758 | 30535990 | ||
| BC112404.1 | - | 1 | 755 | 40060424 | ||

Ndst3
Genomic Map