siRNA Details
| Gene Symbol: Ndst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293682.1 | NP_001280611.1 | 3 | 361 | ||
| NM_031186.3 | NP_112463.2 | 4 | 1474 | |||
| GenBank mRNA | AF221095.1 | - | 4 | 1473 | ||
| AK014990.1 | - | 2 | 514 | |||
| AK015768.1 | - | 3 | 361 | |||
| AK141945.1 | - | 5 | 1623 | |||
| BC079622.1 | - | 4 | 1389 | 30535990 | ||
| BC112404.1 | - | 4 | 1386 | 40060424 | ||

Ndst3
Genomic Map