siRNA Details
| Gene Symbol: Ctns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031251.4 | NP_112541.1 | 4, 5 | 335 | ||
| GenBank mRNA | AJ250670.1 | - | 3, 4 | 233 | ||
| AK054467.1 | - | 5, 6 | 513 | |||
| AK078511.1 | - | 4, 5 | 347 | |||
| AK079295.1 | - | 4, 5 | 336 | |||
| AK137143.1 | - | 4, 5 | 378 | |||
| AK170790.1 | - | 4, 5 | 347 | |||
| AK170828.1 | - | 4, 5 | 358 | |||
| BC005479.1 | - | 4, 5 | 322 | 3484952 | ||
| BC006033.1 | - | 4, 5 | 322 | |||

Ctns
Genomic Map