siRNA Details
| Gene Symbol: Chrdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114385.1 | NP_001107857.1 | 8 | 1161 | ||
| NM_031258.3 | NP_112548.2 | 8 | 1161 | |||
| GenBank mRNA | AF296451.1 | - | 8 | 1083 | ||
| AF321853.1 | - | 7 | 637 | |||
| AK142805.1 | - | 8 | 785 | |||
| AK164661.1 | - | 8 | 756 | |||

Chrdl1
Genomic Map