siRNA Details
| Gene Symbol: Tex16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031382.2 | NP_113559.2 | 2 | 873 | ||
| GenBank mRNA | AF285573.1 | - | 2 | 873 | ||
| BC132332.1 | - | 2 | 897 | 40130609 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex16| Gene Symbol: Tex16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031382.2 | NP_113559.2 | 2 | 873 | ||
| GenBank mRNA | AF285573.1 | - | 2 | 873 | ||
| BC132332.1 | - | 2 | 897 | 40130609 | ||