siRNA Details
| Gene Symbol: Tex11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167997.1 | NP_001161469.1 | 11 | 918 | ||
| NM_031384.2 | NP_113561.2 | 11 | 918 | |||
| GenBank mRNA | AF285572.1 | - | 11 | 916 | ||
| AK029799.1 | - | 11 | 919 | |||
| AK082794.1 | - | 11 | 910 | |||
| BC116709.1 | - | 11 | 914 | 40096708 | ||

Tex11
Genomic Map