siRNA Details
| Gene Symbol: Usp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031388.2 | NP_113565.2 | 8 | 2300 | ||
| GenBank mRNA | AF285570.1 | - | 5 | 1896 | ||
| BC125454.1 | - | 3 | 1799 | 40129969 | ||
| BC132154.1 | - | 3 | 1799 | 40130431 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp26| Gene Symbol: Usp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031388.2 | NP_113565.2 | 8 | 2300 | ||
| GenBank mRNA | AF285570.1 | - | 5 | 1896 | ||
| BC125454.1 | - | 3 | 1799 | 40129969 | ||
| BC132154.1 | - | 3 | 1799 | 40130431 | ||