siRNA Details
Gene Symbol: Wdr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031392.2 | NP_113569.1 | 5 | 2889 | ||
GenBank mRNA | AB041854.1 | - | 5 | 2955 | ||
AF348591.1 | - | 5 | 2885 | |||
AK037181.1 | - | 5 | 2866 | |||
AK046119.1 | - | 5 | 2857 | |||
AK052235.1 | - | 5 | 2830 | |||
AK146617.1 | - | 5 | 2846 | |||
AK163752.1 | - | 5 | 2822 | |||
AK165935.1 | - | 5 | 2853 | |||
AK166265.1 | - | 5 | 2867 | |||
AK169357.1 | - | 5 | 2890 | |||
AK177316.1 | - | 2 | 69 | |||
AK177658.1 | - | 2 | 70 | |||
AK187031.1 | - | 2 | 70 | |||
AK215607.1 | - | 2 | 70 | |||
BC035951.1 | - | 4 | 868 | |||
BC050894.1 | - | 5 | 2889 | 5708880 | ||
BC054367.1 | - | 5 | 2817 | 5346943 |
