siRNA Details
| Gene Symbol: Srrt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109909.1 | NP_001103379.1 | 7 | 1168 | ||
| NM_001109910.1 | NP_001103380.1 | 7 | 1168 | |||
| NM_031405.2 | NP_113582.1 | 7 | 1168 | |||
| GenBank mRNA | AK144300.1 | - | 7 | 1159 | ||
| BC066831.1 | - | 7 | 1127 | 30471339 | ||

Srrt
Genomic Map