siRNA Details
| Gene Symbol: Phip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081216.1 | NP_001074685.1 | 13 | 1368 | ||
| GenBank mRNA | AB049460.1 | - | 13 | 1287 | ||
| AJ303103.1 | - | 13 | 1264 | |||
| AK032797.1 | - | 13 | 1398 | |||
| AK042284.1 | - | 10 | 1534 | |||
| AK049736.1 | - | 13 | 1304 | |||
| AK050025.1 | - | 13 | 1313 | |||
| AK136530.1 | - | 13 | 1306 | |||
| BC009146.1 | - | 4 | 242 | |||
| BC022102.1 | - | 13 | 1309 | |||
| BC037036.1 | - | 13 | 1297 | |||
| BC039765.1 | - | 13 | 1320 | |||
| BC049950.1 | - | 13 | 1298 | |||
| BC068022.1 | - | 4 | 296 | |||

Phip
Genomic Map