siRNA Details
| Gene Symbol: Slmap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032008.3 | NP_114397.3 | 19 | 2205 | ||
| GenBank mRNA | AK129403.1 | - | 19 | 2205 | ||
| AK141597.1 | - | 20 | 3425 | |||
| AK146685.1 | - | 7 | 719 | |||
| BC021457.1 | - | 9 | 968 | 5064534 | ||

Slmap
Genomic Map