siRNA Details
| Gene Symbol: Sucnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032400.2 | NP_115776.2 | 2 | 182 | ||
| GenBank mRNA | AF295367.1 | - | 2 | 198 | ||
| BB864882.1 | - | 2 | 186 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sucnr1| Gene Symbol: Sucnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032400.2 | NP_115776.2 | 2 | 182 | ||
| GenBank mRNA | AF295367.1 | - | 2 | 198 | ||
| BB864882.1 | - | 2 | 186 | |||
Set Membership: |
|