siRNA Details
| Gene Symbol: Sucnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032400.2 | NP_115776.2 | 2 | 280 | ||
| GenBank mRNA | AF295367.1 | - | 2 | 296 | ||
| BB864882.1 | - | 2 | 284 | |||
| BC096665.1 | - | 2 | 253 | 1365839 | ||
More Information
Set Membership: |
|

Sucnr1
Genomic Map