siRNA Details
| Gene Symbol: Rnf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032543.2 | NP_115932.1 | 3, 4 | 369 | ||
| GenBank mRNA | AK137599.1 | - | 3, 4 | 314 | ||
| AK157870.1 | - | 3, 4 | 279 | |||
| AK158320.1 | - | 3, 4 | 321 | |||
| AK172244.1 | - | 3, 4 | 293 | |||
| AY744153.1 | - | 2, 3 | 168 | |||
| BC025530.1 | - | 3, 4 | 290 | |||
| BC057082.1 | - | 3, 4 | 764 | 5704926 | ||
| BY286344.1 | - | 3, 4 | 296 | |||

Rnf123
Genomic Map