siRNA Details
| Gene Symbol: Rnf123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032543.2 | NP_115932.1 | 7 | 663 | ||
| GenBank mRNA | AK137599.1 | - | 7 | 608 | ||
| AK157870.1 | - | 7 | 573 | |||
| AK158320.1 | - | 7 | 615 | |||
| AK172244.1 | - | 7 | 587 | |||
| AK177373.1 | - | 3 | 193 | |||
| AY744153.1 | - | 6 | 462 | |||
| BC025530.1 | - | 7 | 584 | |||
| BC057082.1 | - | 7 | 1058 | 5704926 | ||

Rnf123
Genomic Map