siRNA Details
| Gene Symbol: Sec16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159986.1 | NP_001153458.1 | 11, 12 | 1543 | ||
| NM_033354.3 | NP_203505.3 | 13, 14 | 1790 | |||
| GenBank mRNA | AB063356.1 | - | 11, 12 | 1543 | ||
| AK132144.1 | - | 11, 12 | 1567 | |||
| AK173302.1 | - | 13, 14 | 1771 | |||
| BC039223.1 | - | 3, 4 | 394 | |||
| BC051110.1 | - | 4, 5 | 490 | |||
| BC059194.1 | - | 12, 13 | 1608 | 4222915 | ||

Sec16b
Genomic Map