siRNA Details
| Gene Symbol: Klf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033563.2 | NP_291041.2 | 1 | 401 | ||
| GenBank mRNA | AB221646.1 | - | 1 | 13 | ||
| AF327568.1 | - | 1 | 292 | |||
| AF338369.1 | - | 1 | 436 | |||
| AK149362.1 | - | 2 | 192 | |||
| BC004700.1 | - | 2 | 166 | 3499191 | ||
| BC019604.1 | - | 1 | 401 | 3601534 | ||
| BC040794.1 | - | 2 | 166 | 3499191 | ||

Klf7
Genomic Map