siRNA Details
| Gene Symbol: Pcdhgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033575.3 | NP_291053.1 | 1 | 1397 | ||
| GenBank mRNA | AY013805.1 | - | 1 | 1298 | ||
| BC054557.1 | - | 1 | 1397 | 5706538 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhgb2| Gene Symbol: Pcdhgb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033575.3 | NP_291053.1 | 1 | 1397 | ||
| GenBank mRNA | AY013805.1 | - | 1 | 1298 | ||
| BC054557.1 | - | 1 | 1397 | 5706538 | ||