siRNA Details
| Gene Symbol: Pcdhgb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033577.1 | NP_291055.1 | 1 | 863 | ||
| GenBank mRNA | AY013807.1 | - | 1 | 863 | ||
| BC057403.1 | - | 1 | 1006 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhgb5| Gene Symbol: Pcdhgb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033577.1 | NP_291055.1 | 1 | 863 | ||
| GenBank mRNA | AY013807.1 | - | 1 | 863 | ||
| BC057403.1 | - | 1 | 1006 | |||