siRNA Details
| Gene Symbol: Pcdhgb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033578.3 | NP_291056.1 | 1 | 675 | ||
| GenBank mRNA | AY013808.1 | - | 1 | 528 | ||
| BC054556.1 | - | 1 | 675 | 5702016 | ||
| Gene Symbol: Pcdhga12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF464165.1 | - | 94 | |||

Pcdhgb6
Genomic Map