siRNA Details
| Gene Symbol: Smarca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290708.1 | NP_001277637.1 | 11 | 1651 | ||
| NM_053123.4 | NP_444353.3 | 11 | 1651 | |||
| GenBank mRNA | AF325920.1 | - | 11 | 1658 | ||
| AK030741.1 | - | 11 | 1618 | |||
| AK032978.1 | - | 11 | 1620 | |||
| AK052092.1 | - | 11 | 1607 | |||
| BC016071.1 | - | 11 | 1628 | |||
| BC057115.1 | - | 11 | 1651 | 6406330 | ||

Smarca1
Genomic Map