siRNA Details
| Gene Symbol: Vangl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033509.3 | NP_277044.1 | 4 | 699 | ||
| GenBank mRNA | AF365875.1 | - | 4 | 463 | ||
| AK033128.1 | - | 4 | 617 | |||
| AK046036.1 | - | 4 | 718 | |||
| AK049613.1 | - | 714 | ||||
| AK129311.1 | - | 699 | ||||
| AY035370.3 | - | 4 | 687 | |||
| BC052195.1 | - | 4 | 699 | 6509008 | ||
| BC075636.1 | - | 1556 | ||||

Vangl2
Genomic Map