siRNA Details
| Gene Symbol: Igsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145800.1 | NP_001139272.1 | 19 | 3114 | ||
| NM_033608.3 | NP_291086.2 | 19 | 3180 | |||
| GenBank mRNA | AF317839.1 | - | 19 | 3180 | ||
| AK078536.1 | - | 19 | 3109 | |||
| BC034594.1 | - | 18 | 2943 | |||
| BC040281.1 | - | 19 | 3140 | 5364073 | ||

Igsf9
Genomic Map