siRNA Details
| Gene Symbol: Igsf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145800.1 | NP_001139272.1 | 21 | 3697 | ||
| NM_033608.3 | NP_291086.2 | 21 | 3763 | |||
| GenBank mRNA | AF317839.1 | - | 21 | 3763 | ||
| AK078536.1 | - | 21 | 3693 | |||
| AK145121.1 | - | 21 | 3677 | |||
| BC023011.1 | - | 3 | 557 | |||
| BC024487.1 | - | 3 | 535 | |||
| BC034594.1 | - | 20 | 3526 | |||
| BC040281.1 | - | 21 | 3723 | 5364073 | ||

Igsf9
Genomic Map