siRNA Details
| Gene Symbol: Pnck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199351.1 | NP_001186280.1 | 10 | 1027 | ||
| NM_001199352.1 | NP_001186281.1 | 10 | 891 | |||
| NM_012040.3 | NP_036170.1 | 10 | 967 | |||
| GenBank mRNA | AB023027.1 | - | 9 | 837 | ||
| AF181984.1 | - | 10 | 941 | |||
| AK134665.1 | - | 10 | 898 | |||
| AK158755.1 | - | 10 | 892 | |||
| BC051996.1 | - | 10 | 874 | |||
| BC055891.1 | - | 10 | 867 | 4208888 | ||

Pnck
Genomic Map