siRNA Details
| Gene Symbol: Pcdhb15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053140.3 | NP_444370.1 | 1 | 1329 | ||
| GenBank mRNA | AK038663.1 | - | 1 | 1331 | ||
| AY013776.1 | - | 1 | 1158 | |||
| BC066178.1 | - | 1 | 1335 | 30100073 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhb15| Gene Symbol: Pcdhb15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053140.3 | NP_444370.1 | 1 | 1329 | ||
| GenBank mRNA | AK038663.1 | - | 1 | 1331 | ||
| AY013776.1 | - | 1 | 1158 | |||
| BC066178.1 | - | 1 | 1335 | 30100073 | ||