siRNA Details
Gene Symbol: Glp2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175681.3 | NP_783612.2 | 8 | 1108 | ||
GenBank mRNA | AK035702.1 | - | 503 | |||
AY605231.1 | - | 8 | 827 | |||
BC044746.2 | - | 8 | 1108 | 5363416 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
