siRNA Details
| Gene Symbol: Nkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163660.1 | NP_001157132.1 | 3 | 262 | ||
| NM_027280.3 | NP_081556.3 | 5 | 472 | |||
| GenBank mRNA | AF343352.1 | - | 5 | 309 | ||
| AF358134.1 | - | 5 | 388 | |||
| AK082367.1 | - | 5 | 460 | |||
| BC034838.1 | - | 5 | 446 | 1429943 | ||
| DV662736.1 | - | 3 | 242 | |||

Nkd1
Genomic Map