siRNA Details
| Gene Symbol: Nkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163660.1 | NP_001157132.1 | 8 | 1159 | ||
| NM_027280.3 | NP_081556.3 | 10 | 1369 | |||
| GenBank mRNA | AF343352.1 | - | 10 | 1206 | ||
| AF358134.1 | - | 10 | 1285 | |||
| AK076164.1 | - | 10 | 1360 | |||
| AK082367.1 | - | 10 | 1358 | |||
| BC034838.1 | - | 10 | 1343 | 1429943 | ||

Nkd1
Genomic Map