siRNA Details
| Gene Symbol: Hemgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053149.2 | NP_444379.1 | 1, 2 | 267 | ||
| GenBank mRNA | AF269248.1 | - | 1, 2 | 267 | ||
| AK006588.1 | - | 2, 3 | 282 | |||
| AK076587.1 | - | 2, 3 | 252 | |||
| AK077692.1 | - | 1, 2 | 110 | |||
| AK156536.1 | - | 1, 2 | 195 | |||
| AK171890.1 | - | 1, 2 | 186 | |||
| AK172421.1 | - | 1, 2 | 186 | |||
| AY699986.1 | - | 1, 2 | 109 | |||
| BC050798.1 | - | 2, 3 | 246 | 6743819 | ||

Hemgn
Genomic Map