siRNA Details
| Gene Symbol: Clmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040682.1 | NP_001035772.1 | 9 | 1537 | ||
| NM_053155.2 | NP_444385.2 | 9 | 1537 | |||
| GenBank mRNA | AB047978.1 | - | 9 | 1530 | ||
| AB059643.1 | - | 9 | 1530 | |||
| AB059644.1 | - | 9 | 1530 | |||
| AB059645.1 | - | 9 | 1530 | |||
| AB059646.1 | - | 9 | 1530 | |||
| AB059647.1 | - | 9 | 1530 | |||
| AB059648.1 | - | 9 | 1530 | |||
| AK077023.1 | - | 6 | 1119 | |||
| AK147777.1 | - | 9 | 1434 | |||
| AK162015.1 | - | 1 | 1254 | |||
| BC019176.1 | - | 9 | 1468 | |||
| BC096470.1 | - | 9 | 1426 | |||
| BC141280.1 | - | 9 | 1470 | 9055892 | ||

Clmn
Genomic Map