siRNA Details
Gene Symbol: Tm2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053157.2 | NP_444387.1 | 4, 5 | 466 | ||
GenBank mRNA | AF353993.1 | - | 4, 5 | 440 | ||
AK009511.1 | - | 4, 5 | 430 | |||
AK216092.1 | - | 4, 5 | 344 | |||
BC089492.1 | - | 4, 5 | 466 | 6703812 |
