siRNA Details
| Gene Symbol: Tm2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053157.2 | NP_444387.1 | 3, 4 | 380 | ||
| GenBank mRNA | AF353993.1 | - | 3, 4 | 354 | ||
| AK009511.1 | - | 3, 4 | 344 | |||
| AK216092.1 | - | 3, 4 | 258 | |||
| BC089492.1 | - | 3, 4 | 380 | 6703812 | ||

Tm2d1
Genomic Map