siRNA Details
| Gene Symbol: Hrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053176.2 | NP_444406.2 | 7 | 915 | ||
| GenBank mRNA | AB055897.1 | - | 7 | 873 | ||
| AF194028.1 | - | 7 | 855 | |||
| AY135662.1 | - | 7 | 876 | |||
| BC011168.1 | - | 7 | 861 | 4195766 | ||
| CB946752.1 | - | 3 | 263 | |||

Hrg
Genomic Map