siRNA Details
| Gene Symbol: Ddx50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053183.2 | NP_444413.1 | 5, 6 | 824 | ||
| GenBank mRNA | AF334104.1 | - | 5, 6 | 825 | ||
| AK030239.1 | - | 6, 7 | 896 | |||
| AK041281.1 | - | 5, 6 | 723 | |||
| BC049906.1 | - | 5, 6 | 825 | |||
| BC055440.1 | - | 5, 6 | 818 | |||
| BC075714.1 | - | 5, 6 | 776 | |||
| BC090996.1 | - | 5, 6 | 844 | 6757408 | ||

Ddx50
Genomic Map