siRNA Details
Gene Symbol: Cnnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039551.1 | NP_001034640.1 | 2, 3 | 1414 | ||
NM_053186.2 | NP_444416.2 | 2, 3 | 1414 | |||
GenBank mRNA | AF216964.1 | - | 2, 3 | 483 | ||
AK143035.1 | - | 1, 2 | 116 | |||
BC080275.1 | - | 2, 3 | 841 | |||
BC108417.1 | - | 2, 3 | 668 | |||
BC132285.1 | - | 2, 3 | 466 | |||
BC144720.1 | - | 2, 3 | 465 | |||
GQ225746.2 | - | 2, 3 | 400 |
