siRNA Details
| Gene Symbol: Cnnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102471.1 | NP_001095941.1 | 3, 4 | 2067 | ||
| NM_033569.3 | NP_291047.2 | 3, 4 | 2067 | |||
| GenBank mRNA | AF216961.1 | - | 3, 4 | 1379 | ||
| AK032006.1 | - | 3, 4 | 2026 | |||
| AK053711.1 | - | 3, 4 | 2085 | |||
| AK159616.1 | - | 3, 4 | 2073 | |||
| BC052513.1 | - | 1, 2 | 82 | |||
| BC125033.1 | - | 3, 4 | 1379 | |||
| GQ225745.1 | - | 3, 4 | 1366 | |||

Cnnm2
Genomic Map