siRNA Details
| Gene Symbol: Cnnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102471.1 | NP_001095941.1 | 5 | 2278 | ||
| NM_033569.3 | NP_291047.2 | 5 | 2278 | |||
| GenBank mRNA | AF216961.1 | - | 5 | 1590 | ||
| AK032006.1 | - | 5 | 2237 | |||
| AK053711.1 | - | 5 | 2296 | |||
| AK159616.1 | - | 5 | 2284 | |||
| BC038027.1 | - | 170 | ||||
| BC052513.1 | - | 3 | 293 | |||
| BC125033.1 | - | 5 | 1590 | |||
| GQ225745.1 | - | 5 | 1577 | |||

Cnnm2
Genomic Map