siRNA Details
| Gene Symbol: Cnnm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033570.2 | NP_291048.2 | 1 | 642 | ||
| GenBank mRNA | AF216963.1 | - | 1 | 254 | ||
| AK173209.1 | - | 1 | 619 | |||
| BC139799.1 | - | 1 | 243 | |||
| GU390541.1 | - | 1 | 177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnnm4| Gene Symbol: Cnnm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033570.2 | NP_291048.2 | 1 | 642 | ||
| GenBank mRNA | AF216963.1 | - | 1 | 254 | ||
| AK173209.1 | - | 1 | 619 | |||
| BC139799.1 | - | 1 | 243 | |||
| GU390541.1 | - | 1 | 177 | |||