siRNA Details
| Gene Symbol: Dgcr8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033324.2 | NP_201581.2 | 4, 5 | 1363 | ||
| GenBank mRNA | AB086857.1 | - | 4, 5 | 1363 | ||
| AK048056.1 | - | 3, 4 | 1164 | |||
| BC051013.1 | - | 2, 3 | 1549 | |||
| BC062919.1 | - | 4, 5 | 1383 | 6833280 | ||
| BC066118.1 | - | 4, 5 | 1410 | 6400561 | ||

Dgcr8
Genomic Map