siRNA Details
| Gene Symbol: Pi15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053191.2 | NP_444421.2 | 2 | 228 | ||
| GenBank mRNA | AF329196.1 | - | 2 | 148 | ||
| AK030960.1 | - | 2 | 229 | |||
| AK040961.1 | - | 2 | 227 | |||
| BC116705.1 | - | 2 | 124 | 40102877 | ||

Pi15
Genomic Map