siRNA Details
| Gene Symbol: Tinagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168333.1 | NP_001161805.1 | 7 | 807 | ||
| NM_023476.3 | NP_075965.2 | 7 | 948 | |||
| GenBank mRNA | AB047402.1 | - | 7 | 949 | ||
| AB050626.1 | - | 7 | 785 | |||
| AK087142.1 | - | 808 | ||||
| BC005738.1 | - | 7 | 775 | 2650851 | ||
| BC018539.1 | - | 7 | 792 | 3596560 | ||
| CT010275.1 | - | 6 | 566 | |||

Tinagl1
Genomic Map