siRNA Details
| Gene Symbol: Loxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033325.2 | NP_201582.2 | 3 | 646 | ||
| GenBank mRNA | AK034957.1 | - | 3 | 684 | ||
| AK045019.1 | - | 3 | 646 | |||
| AK145323.1 | - | 3 | 684 | |||
| AK159386.1 | - | 3 | 680 | |||
| BC086801.1 | - | 782 | 30605240 | |||

Loxl2
Genomic Map