siRNA Details
| Gene Symbol: Susd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144796.4 | NP_659045.2 | 5 | 881 | ||
| GenBank mRNA | AK035779.1 | - | 5 | 881 | ||
| AK088629.1 | - | 5 | 794 | |||
| BC021842.1 | - | 5 | 897 | 5135798 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Susd4| Gene Symbol: Susd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144796.4 | NP_659045.2 | 5 | 881 | ||
| GenBank mRNA | AK035779.1 | - | 5 | 881 | ||
| AK088629.1 | - | 5 | 794 | |||
| BC021842.1 | - | 5 | 897 | 5135798 | ||