siRNA Details
| Gene Symbol: Deptor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037937.3 | NP_001033026.1 | 3 | 323 | ||
| NM_145470.3 | NP_663445.2 | 3 | 501 | |||
| GenBank mRNA | AK139888.1 | - | 3 | 323 | ||
| AK145208.1 | - | 3 | 395 | |||
| AK220300.1 | - | 3 | 335 | |||
| BC004774.1 | - | 3 | 506 | |||
| DV649395.1 | - | 3 | 468 | |||

Deptor
Genomic Map