siRNA Details
| Gene Symbol: Eif3m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145380.2 | NP_663355.1 | 11 | 1090 | ||
| GenBank mRNA | AK077480.1 | - | 11 | 1091 | ||
| AK084738.1 | - | 11 | 1109 | |||
| AK146222.1 | - | 11 | 1122 | |||
| AK168066.1 | - | 11 | 1107 | |||
| BC005598.1 | - | 11 | 1090 | 3969335 | ||
| BC050191.1 | - | 5 | 1822 | |||
| BC103795.1 | - | 11 | 1090 | 30135815 | ||
| BC116787.1 | - | 11 | 1087 | 40126106 | ||
| BC116789.1 | - | 11 | 1087 | 40126108 | ||

Eif3m
Genomic Map