siRNA Details
| Gene Symbol: Lrrc59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133807.1 | NP_598568.1 | 4 | 571 | ||
| GenBank mRNA | AK053450.1 | - | 4 | 598 | ||
| AK159374.1 | - | 4 | 627 | |||
| AK159552.1 | - | 4 | 621 | |||
| AK166576.1 | - | 4 | 620 | |||
| AK167604.1 | - | 4 | 562 | |||
| AK169250.1 | - | 4 | 546 | |||
| AK169390.1 | - | 4 | 570 | |||
| AK196953.1 | - | 144 | ||||
| BC006877.1 | - | 4 | 571 | 2647513 | ||
| BC066172.1 | - | 4 | 565 | 30429192 | ||

Lrrc59
Genomic Map