siRNA Details
| Gene Symbol: Kmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133809.1 | NP_598570.1 | 15 | 1299 | ||
| GenBank mRNA | AK129011.1 | - | 1210 | |||
| AK133074.1 | - | 1221 | ||||
| AK171812.1 | - | 15 | 1427 | |||
| BC014683.1 | - | 15 | 1299 | 4210278 | ||
| BC024455.1 | - | 4 | 251 | |||

Kmo
Genomic Map