siRNA Details
| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133810.3 | NP_598571.2 | 4 | 566 | ||
| GenBank mRNA | AK029551.1 | - | 4 | 507 | ||
| AK035693.1 | - | 4 | 534 | |||
| AK076932.1 | - | 4 | 533 | |||
| AK083332.1 | - | 4 | 535 | |||
| AK153108.1 | - | 4 | 510 | |||
| AK165291.1 | - | 4 | 506 | |||
| AY092028.1 | - | 4 | 519 | |||
| BC006579.1 | - | 4 | 521 | 3586767 | ||

Stk17b
Genomic Map