siRNA Details
| Gene Symbol: Gorab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178883.5 | NP_849214.2 | 3, 4 | 569 | ||
| GenBank mRNA | AK039170.1 | - | 3, 4 | 580 | ||
| AK043953.1 | - | 3, 4 | 570 | |||
| AK165909.1 | - | 3, 4 | 539 | |||
| AK169151.1 | - | 3, 4 | 562 | |||
| BC119132.1 | - | 3, 4 | 552 | 8733881 | ||
| BC119134.1 | - | 3, 4 | 552 | 8733883 | ||

Gorab
Genomic Map